Gene Transcripts
- Gene Symbol
- RMST
- Entrez Gene ID
- 196475
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-protein coding)
- Gene Aliases
- LINC00054, NCRMS, NCRNA00054
- Chromosome Location
- Chr. 12: 97465021 - 97533766 on Build GRCh38
- UniGene ID
- Hs.652568
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024037.1 |
|
7 |
752 |
|
GenBank mRNA |
AF429305.1 |
- |
5 |
685 |
|
AK056164.1 |
- |
7 |
756 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA746072.1 |
- |
3 |
315 |
|
Chr. 12 - 97493950 - 97530844 [+] |
GenBankmRNA |
DA746073.1 |
- |
2 |
317 |
|
Chr. 12 - 97494742 - 97530844 [+] |
View Full Product Details