Gene Transcripts

Gene Symbol
MIR155HG
Entrez Gene ID
114614
Gene Name
MIR155 host gene
Gene Aliases
BIC, MIRHG2, NCRNA00172
Chromosome Location
Chr. 21: 25562145 - 25575168 on Build GRCh38
UniGene ID
Hs.697120
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001458.3
3 840
GenBank mRNA AF402776.1 - LIT1586
bic
3 792

View Full Product Details