Gene Transcripts

Gene Symbol
SNHG5
Entrez Gene ID
387066
Gene Name
small nucleolar RNA host gene 5
Gene Aliases
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
Chromosome Location
Chr. 6: 85677007 - 85678733 on Build GRCh38
UniGene ID
Hs.292457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003038.2 6 342
GenBank mRNA BC009220.2 - 5 816
BC108291.1 - 5 834
CD384894.1 - 6 359
CD388674.1 - 6 359


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD389879.1 - 6 365 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD173365.1 - 6 368 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA DA482879.1 - 6 342 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD520230.1 - 6 345 Chr. 6 - 85677083 - 85678736 [-]
GenBankmRNA BU532076.1 - 345 Chr. - - [-]
GenBankmRNA BE715636.1 - 5 309 Chr. 6 - 85677108 - 85678173 [-]
GenBankmRNA BE931917.1 - 5 483 Chr. 6 - 85677155 - 85678356 [-]
GenBankmRNA CD247680.1 - 6 362 Chr. 6 - 85677082 - 85678724 [-]
GenBankmRNA AW972834.1 - 436 Chr. - - [-]
GenBankmRNA CD249906.1 - 6 348 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA CD389769.1 - 6 361 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD175213.1 - 6 358 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA CN368113.1 - 5 321 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA BU536948.1 - 340 Chr. - - [-]
GenBankmRNA CD048947.1 - 369 Chr. - - [-]
GenBankmRNA BU500478.1 - 6 325 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA BE392169.1 - 5 299 Chr. 6 - 85677079 - 85678185 [-]
GenBankmRNA CD388186.1 - 6 363 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD248605.1 - 6 360 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD177483.1 - 6 358 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA CN368120.1 - 6 334 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA CD175092.1 - 6 365 Chr. 6 - 85677082 - 85678724 [-]
GenBankmRNA CD249877.1 - 6 365 Chr. 6 - 85677079 - 85678725 [-]
GenBankmRNA CD385054.1 - 369 Chr. - - [-]
GenBankmRNA CD250044.1 - 6 370 Chr. 6 - 85677082 - 85678734 [-]
GenBankmRNA CD049899.1 - 360 Chr. - - [-]
GenBankmRNA CD248963.1 - 364 Chr. - - [-]
GenBankmRNA BM452606.1 - 5 301 Chr. 6 - 85677079 - 85678186 [-]
GenBankmRNA CD389793.1 - 363 Chr. - - [-]
GenBankmRNA BF966370.2 - 6 348 Chr. 6 - 85677082 - 85678737 [-]
GenBankmRNA BU526489.1 - 6 317 Chr. 6 - 85677082 - 85678709 [-]
GenBankmRNA N98867.1 - 5 338 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA BG330935.1 - 6 333 Chr. 6 - 85677079 - 85678723 [-]
GenBankmRNA BU588037.1 - 345 Chr. - - [-]
GenBankmRNA BF980513.1 - 319 Chr. - - [-]
GenBankmRNA CB117414.1 - 6 321 Chr. 6 - 85677082 - 85678712 [-]
GenBankmRNA BU191449.1 - 6 323 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BG705462.1 - 6 348 Chr. 6 - 85677082 - 85678737 [-]
GenBankmRNA AA308290.1 - 6 318 Chr. 6 - 85677083 - 85678709 [-]
GenBankmRNA AJ714332.1 - 6 335 Chr. 6 - 85677100 - 85678715 [-]
GenBankmRNA CD177664.1 - 6 362 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA BU602968.1 - 346 Chr. - - [-]
GenBankmRNA CD172948.1 - 6 361 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA HY027691.1 - 6 341 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA BU181781.1 - 319 Chr. - - [-]
GenBankmRNA CD174861.1 - 6 365 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BU941995.1 - 345 Chr. - - [-]
GenBankmRNA BQ896708.1 - 6 321 Chr. 6 - 85677082 - 85678713 [-]
GenBankmRNA BG758157.1 - 6 327 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA CD174362.1 - 6 366 Chr. 6 - 85677082 - 85678734 [-]
GenBankmRNA BU536398.1 - 6 345 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA CD172704.1 - 6 362 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA F25212.1 - 6 325 Chr. 6 - 85677105 - 85678716 [-]
GenBankmRNA BF663376.1 - 325 Chr. - - [-]
GenBankmRNA CD175429.1 - 6 360 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA BU931690.1 - 5 285 Chr. 6 - 85677082 - 85678171 [-]
GenBankmRNA CD250358.1 - 1 368 Chr. 17 - 49339407 - 49339817 [-]
GenBankmRNA BG760195.1 - 6 326 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA BE736061.1 - 5 307 Chr. 6 - 85677079 - 85678192 [-]
GenBankmRNA BF309660.1 - 5 314 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA BU169141.1 - 6 324 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BM476100.1 - 331 Chr. - - [-]
GenBankmRNA BM469398.1 - 5 302 Chr. 6 - 85677079 - 85678188 [-]
GenBankmRNA HY104298.1 - 6 343 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA BU600470.1 - 6 348 Chr. 6 - 85677077 - 85678736 [-]
GenBankmRNA F25999.1 - 6 326 Chr. 6 - 85677082 - 85678717 [-]
GenBankmRNA CD174641.1 - 6 364 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD246809.1 - 6 358 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA CB996821.1 - 6 365 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BG398388.1 - 319 Chr. - - [-]
GenBankmRNA BE257562.1 - 5 306 Chr. 6 - 85677081 - 85678192 [-]
GenBankmRNA CD000147.1 - 346 Chr. - - [-]
GenBankmRNA CD676465.1 - 5 385 Chr. 6 - 85677082 - 85678252 [-]
GenBankmRNA BU532819.1 - 6 345 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA BG754322.1 - 326 Chr. - - [-]
GenBankmRNA BG419815.1 - 6 336 Chr. 6 - 85677077 - 85678726 [-]
GenBankmRNA BG332320.1 - 6 325 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA HY067002.1 - 6 341 Chr. 6 - 85677082 - 85678732 [-]
GenBankmRNA CD174976.1 - 6 360 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA F33656.1 - 296 Chr. - - [-]
GenBankmRNA CD387937.1 - 359 Chr. - - [-]
GenBankmRNA HX785786.1 - 6 698 Chr. 6 - 85677147 - 85678602 [-]
GenBankmRNA AV712595.2 - 6 342 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BG759432.1 - 6 319 Chr. 6 - 85677082 - 85678710 [-]
GenBankmRNA CD250171.1 - 6 360 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA HY181073.1 - 6 342 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA CD249518.1 - 6 368 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA HY031639.1 - 6 341 Chr. 6 - 85677084 - 85678732 [-]
GenBankmRNA F24362.1 - 5 298 Chr. 6 - 85677119 - 85678184 [-]
GenBankmRNA CD177169.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA F19304.2 - 326 Chr. 11 - 8244443 - 8244457 [+]
GenBankmRNA BM920267.1 - 326 Chr. - - [-]
GenBankmRNA EB388820.1 - 6 331 Chr. 6 - 85677082 - 85678722 [-]
GenBankmRNA CN368117.1 - 6 334 Chr. 6 - 85677007 - 85678717 [-]
GenBankmRNA BQ432969.1 - 6 351 Chr. 6 - 85677082 - 85678719 [-]
GenBankmRNA CD385963.1 - 6 364 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA F31772.1 - 5 299 Chr. 6 - 85677119 - 85678185 [-]
GenBankmRNA CD174499.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CF780557.1 - 6 325 Chr. 6 - 85677079 - 85678716 [-]
GenBankmRNA BU182289.1 - 6 328 Chr. 6 - 85677082 - 85678719 [-]
GenBankmRNA BM695213.1 - 6 324 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BU531826.1 - 6 345 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA BQ230279.1 - 323 Chr. - - [-]
GenBankmRNA DB449890.1 - 6 346 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA BU954986.1 - 345 Chr. - - [-]
GenBankmRNA BF971639.1 - 326 Chr. - - [-]
GenBankmRNA CD175168.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD639840.1 - 515 Chr. - - [-]
GenBankmRNA CD173866.1 - 6 361 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA HX789156.1 - 6 698 Chr. 6 - 85677147 - 85678602 [-]
GenBankmRNA CD177114.1 - 6 364 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD247795.1 - 6 361 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA BU532664.1 - 6 345 Chr. 6 - 85677085 - 85678731 [-]
GenBankmRNA CD176393.1 - 6 368 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BU845184.1 - 5 315 Chr. 6 - 85677082 - 85678192 [-]
GenBankmRNA CD175058.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA F22587.1 - 303 Chr. - - [-]
GenBankmRNA BU536013.1 - 6 345 Chr. 6 - 85677082 - 85678736 [-]
GenBankmRNA CD388557.1 - 6 361 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD175102.1 - 6 363 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CB962559.1 - 6 345 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA BU196777.1 - 5 313 Chr. 6 - 85677082 - 85678183 [-]
GenBankmRNA BE901509.1 - 301 Chr. - - [-]
GenBankmRNA CB128964.1 - 6 342 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BG759735.1 - 6 325 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA HY168432.1 - 6 342 Chr. 6 - 85677083 - 85678732 [-]
GenBankmRNA BU855495.1 - 6 321 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA BQ645788.1 - 6 323 Chr. 6 - 85677082 - 85678714 [-]
GenBankmRNA AA515848.1 - 6 332 Chr. 6 - 85677082 - 85678714 [-]
GenBankmRNA BM840935.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD248175.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BQ897037.1 - 5 313 Chr. 6 - 85677082 - 85678188 [-]
GenBankmRNA CD174771.1 - 6 362 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA AJ709757.1 - 6 330 Chr. 6 - 85677084 - 85678716 [-]
GenBankmRNA CD173592.1 - 6 367 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD387585.1 - 357 Chr. - - [-]
GenBankmRNA BU185686.1 - 6 323 Chr. 6 - 85677082 - 85678715 [-]
GenBankmRNA BF218089.1 - 6 346 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD173436.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD175427.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD177627.1 - 6 369 Chr. 6 - 85677082 - 85678734 [-]
GenBankmRNA BE513854.1 - 5 302 Chr. 6 - 85677081 - 85678188 [-]
GenBankmRNA F19218.2 - 309 Chr. 1 - 203128824 - 203128850 [-]
GenBankmRNA BE256443.1 - 308 Chr. - - [-]
GenBankmRNA BI090148.1 - 326 Chr. - - [-]
GenBankmRNA CD175562.1 - 6 364 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD176074.1 - 6 368 Chr. 6 - 85677085 - 85678734 [-]
GenBankmRNA AJ574636.1 - 6 333 Chr. 6 - 85677087 - 85678718 [-]
GenBankmRNA CD175083.1 - 6 360 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD640064.1 - 322 Chr. - - [-]
GenBankmRNA BE900923.1 - 5 306 Chr. 6 - 85677084 - 85678192 [-]
GenBankmRNA HY101068.1 - 6 341 Chr. 6 - 85677079 - 85678732 [-]
GenBankmRNA CD048516.1 - 399 Chr. - - [-]
GenBankmRNA CD175206.1 - 6 371 Chr. 6 - 85677077 - 85678733 [-]
GenBankmRNA BG754036.1 - 6 325 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA CD177343.1 - 6 365 Chr. 6 - 85677085 - 85678732 [-]
GenBankmRNA AA314904.1 - 5 303 Chr. 6 - 85677123 - 85678189 [-]
GenBankmRNA BM763888.1 - 6 342 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD675327.1 - 6 338 Chr. 6 - 85677082 - 85678720 [-]
GenBankmRNA AA313726.1 - 5 303 Chr. 6 - 85677077 - 85678189 [-]
GenBankmRNA CD173371.1 - 6 368 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD248801.1 - 6 360 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD248278.1 - 6 367 Chr. 6 - 85677077 - 85678733 [-]
GenBankmRNA CD388259.1 - 6 365 Chr. 6 - 85677075 - 85678732 [-]
GenBankmRNA BG339050.1 - 323 Chr. - - [-]
GenBankmRNA CD390099.1 - 6 357 Chr. 6 - 85677083 - 85678721 [-]
GenBankmRNA N55638.1 - 5 336 Chr. 6 - 85677127 - 85678192 [-]
GenBankmRNA BM772443.1 - 6 342 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA BE812115.1 - 5 324 Chr. 6 - 85677119 - 85678192 [-]
GenBankmRNA F24856.1 - 6 321 Chr. 6 - 85677133 - 85678712 [-]
GenBankmRNA BU536630.1 - 6 345 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD176142.1 - 6 332 Chr. 6 - 85677082 - 85678723 [-]
GenBankmRNA CN368115.1 - 6 339 Chr. 6 - 85677087 - 85678724 [-]
GenBankmRNA BG424974.1 - 6 335 Chr. 6 - 85677084 - 85678726 [-]
GenBankmRNA CB956756.1 - 6 342 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA BG472840.1 - 6 326 Chr. 6 - 85677114 - 85678716 [-]
GenBankmRNA F36060.1 - 279 Chr. - - [-]
GenBankmRNA CD389175.1 - 6 362 Chr. 6 - 85677082 - 85678732 [-]
GenBankmRNA CD172639.1 - 6 363 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA AJ707924.1 - 6 360 Chr. 6 - 85677084 - 85678716 [-]
GenBankmRNA CD248034.1 - 7 367 Chr. 6 - 85677079 - 86545552 [-]
GenBankmRNA AB017710.3 - 1 2188 Chr. 6 - 85677007 - 85679361 [-]
GenBankmRNA BM006651.1 - 6 326 Chr. 6 - 85677082 - 85678716 [-]
GenBankmRNA CD248379.1 - 6 367 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD638710.1 - 6 345 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD390234.1 - 6 367 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA BG685417.1 - 6 320 Chr. 6 - 85677082 - 85678710 [-]
GenBankmRNA CD247900.1 - 6 362 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD390212.1 - 6 355 Chr. 6 - 85677082 - 85678733 [-]
GenBankmRNA CD173118.1 - 6 359 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA HY103231.1 - 6 340 Chr. 6 - 85677082 - 85678731 [-]
GenBankmRNA F37437.1 - 325 Chr. - - [-]
GenBankmRNA AL525144.3 - 5 838 Chr. 6 - 85677103 - 85678710 [-]
GenBankmRNA CD175900.1 - 6 362 Chr. 6 - 85677079 - 85678731 [-]
GenBankmRNA BF974828.1 - 6 320 Chr. 6 - 85677082 - 85678710 [-]
GenBankmRNA CN368114.1 - 6 341 Chr. 6 - 85677087 - 85678720 [-]
GenBankmRNA CD174275.1 - 6 364 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BQ935537.1 - 5 345 Chr. 6 - 85677075 - 85678191 [-]
GenBankmRNA BM841447.1 - 6 363 Chr. 6 - 85677134 - 85678733 [-]
GenBankmRNA AA401334.1 - 5 308 Chr. 6 - 85677047 - 85678192 [-]
GenBankmRNA CD173175.1 - 6 359 Chr. 6 - 85677085 - 85678733 [-]
GenBankmRNA CD174931.1 - 6 359 Chr. 6 - 85677082 - 85678733 [-]

View Full Product Details