Gene Transcripts
- Gene Symbol
- SNHG5
- Entrez Gene ID
- 387066
- Gene Name
- small nucleolar RNA host gene 5
- Gene Aliases
- C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
- Chromosome Location
- Chr. 6: 85677007 - 85678733 on Build GRCh38
- UniGene ID
- Hs.292457
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003038.2 |
|
6 |
468 |
|
GenBank mRNA |
CD384894.1 |
- |
6 |
485 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD172637.1 |
- |
6 |
486 |
|
Chr. 6 - 85677007 - 85678733 [-] |
GenBankmRNA |
CD640064.1 |
- |
|
448 |
|
Chr. - - [-] |
GenBankmRNA |
AB017710.3 |
- |
1 |
2314 |
|
Chr. 6 - 85677007 - 85679361 [-] |
GenBankmRNA |
CN368117.1 |
- |
6 |
460 |
|
Chr. 6 - 85677007 - 85678717 [-] |
GenBankmRNA |
BQ230279.1 |
- |
|
449 |
|
Chr. - - [-] |
View Full Product Details