Gene Transcripts

Gene Symbol
SNHG5
Entrez Gene ID
387066
Gene Name
small nucleolar RNA host gene 5
Gene Aliases
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
Chromosome Location
Chr. 6: 85677007 - 85678733 on Build GRCh38
UniGene ID
Hs.292457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003038.2 6 425
GenBank mRNA BC009220.2 - 5 899
CD384894.1 - 6 442


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD172948.1 - 6 444 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD389879.1 - 6 448 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD176393.1 - 6 451 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD249518.1 - 6 451 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD248034.1 - 7 450 Chr. 6 - 85677079 - 86545552 [-]
GenBankmRNA HY101068.1 - 6 424 Chr. 6 - 85677079 - 85678732 [-]
GenBankmRNA CD640064.1 - 405 Chr. - - [-]
GenBankmRNA BU600470.1 - 6 431 Chr. 6 - 85677077 - 85678736 [-]
GenBankmRNA CD175206.1 - 6 454 Chr. 6 - 85677077 - 85678733 [-]
GenBankmRNA CD249877.1 - 6 448 Chr. 6 - 85677079 - 85678725 [-]
GenBankmRNA CD388557.1 - 6 444 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA AB017710.3 - 1 2271 Chr. 6 - 85677007 - 85679361 [-]
GenBankmRNA CN368117.1 - 6 417 Chr. 6 - 85677007 - 85678717 [-]
GenBankmRNA CD638710.1 - 6 428 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD174641.1 - 6 447 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD175102.1 - 6 446 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD172637.1 - 6 443 Chr. 6 - 85677007 - 85678733 [-]
GenBankmRNA DA482879.1 - 6 425 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CB962559.1 - 6 428 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CF780557.1 - 6 408 Chr. 6 - 85677079 - 85678716 [-]
GenBankmRNA CD173118.1 - 6 442 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA BM452606.1 - 5 384 Chr. 6 - 85677079 - 85678186 [-]
GenBankmRNA BG754322.1 - 409 Chr. - - [-]
GenBankmRNA BG419815.1 - 6 420 Chr. 6 - 85677077 - 85678726 [-]
GenBankmRNA BU941995.1 - 428 Chr. - - [-]
GenBankmRNA AA313726.1 - 5 386 Chr. 6 - 85677077 - 85678189 [-]
GenBankmRNA CD248278.1 - 6 450 Chr. 6 - 85677077 - 85678733 [-]
GenBankmRNA CD388259.1 - 6 448 Chr. 6 - 85677075 - 85678732 [-]
GenBankmRNA BG330935.1 - 6 416 Chr. 6 - 85677079 - 85678723 [-]
GenBankmRNA CD387585.1 - 440 Chr. - - [-]
GenBankmRNA BF218089.1 - 6 429 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD387937.1 - 442 Chr. - - [-]
GenBankmRNA BU536630.1 - 6 428 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA CD175900.1 - 6 445 Chr. 6 - 85677079 - 85678731 [-]
GenBankmRNA BQ935537.1 - 5 428 Chr. 6 - 85677075 - 85678191 [-]
GenBankmRNA CD174275.1 - 6 447 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA AA401334.1 - 5 391 Chr. 6 - 85677047 - 85678192 [-]
GenBankmRNA BE256443.1 - 391 Chr. - - [-]
GenBankmRNA CB956756.1 - 6 425 Chr. 6 - 85677079 - 85678736 [-]
GenBankmRNA BE392169.1 - 5 383 Chr. 6 - 85677079 - 85678185 [-]
GenBankmRNA CD388186.1 - 6 446 Chr. 6 - 85677079 - 85678733 [-]
GenBankmRNA CD177114.1 - 6 447 Chr. 6 - 85677079 - 85678733 [-]

View Full Product Details