Gene Transcripts
A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AJ297963.3 |
- |
1 |
236 |
|
Chr. Y - 8739573 - 8739969 [-] |
View Full Product Details