Gene Transcripts

Gene Symbol
RBMS3
Entrez Gene ID
27303
Gene Name
RNA binding motif single stranded interacting protein 3
Gene Aliases
Chromosome Location
Chr. 3: 29280860 - 30010395 on Build GRCh38
UniGene ID
Hs.221436
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003792.2 NP_001003792.1 14 6095
NM_001003793.2 NP_001003793.1 15 6149
NM_001177711.1 NP_001171182.1 14 6101
XM_005265063.1 XP_005265120.1 12 5806
XM_005265065.4 XP_005265122.1 12 5499
XM_017006179.1 XP_016861668.1 14 6550
XM_017006180.1 XP_016861669.1 13 6502
XM_017006181.1 XP_016861670.1 13 6499
XM_017006182.1 XP_016861671.1 14 6578
GenBank mRNA AK093863.1 - 1 1090

View Full Product Details