Gene Transcripts
- Gene Symbol
- GAS2L3
- Entrez Gene ID
- 283431
- Gene Name
- growth arrest specific 2 like 3
- Gene Aliases
- G2L3
- Chromosome Location
- Chr. 12: 100573661 - 100628288 on Build GRCh38
- UniGene ID
- Hs.20575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX649059.1 |
- |
1 |
1563 |
|
View Full Product Details