Gene Transcripts
- Gene Symbol
- MIB2
- Entrez Gene ID
- 142678
- Gene Name
- mindbomb E3 ubiquitin protein ligase 2
- Gene Aliases
- ZZANK1, ZZZ5
- Chromosome Location
- Chr. 1: 1615188 - 1630610 on Build GRCh38
- UniGene ID
- Hs.135805
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC035078.1 |
- |
1 |
1962 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB547595.1 |
- |
1 |
362 |
|
Chr. 1 - 1613760 - 1614195 [-] |
GenBankmRNA |
CN294875.1 |
- |
1 |
277 |
|
Chr. 1 - 1613758 - 1614110 [-] |
GenBankmRNA |
BI018590.1 |
- |
|
169 |
|
Chr. - - [-] |
GenBankmRNA |
BQ189371.1 |
- |
|
251 |
|
Chr. - - [-] |
GenBankmRNA |
BM695863.1 |
- |
1 |
254 |
|
Chr. 1 - 1613758 - 1614074 [-] |
View Full Product Details