Gene Transcripts

Gene Symbol
G3BP1
Entrez Gene ID
10146
Gene Name
G3BP stress granule assembly factor 1
Gene Aliases
G3BP, HDH-VIII
Chromosome Location
Chr. 5: 151771915 - 151811352 on Build GRCh38
UniGene ID
Hs.3353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006714749.3 XP_006714812.1 12 5270
XM_006714750.3 XP_006714813.1 12 5209
XM_017008922.1 XP_016864411.1 12 5556
XM_017008923.1 XP_016864412.1 12 5525
GenBank mRNA AK128316.1 - 1 1047

View Full Product Details