Gene Transcripts

Gene Symbol
SNRPD3
Entrez Gene ID
6634
Gene Name
small nuclear ribonucleoprotein D3 polypeptide
Gene Aliases
SMD3, Sm-D3
Chromosome Location
Chr. 22: 24555650 - 24574964 on Build GRCh38
UniGene ID
Hs.356549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278656.1 NP_001265585.1 4 3029
NM_004175.4 NP_004166.1 4 3043
NR_103819.1 3 2850


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI822977.1 - 1 64 Chr. 22 - 24574137 - 24574430 [+]
GenBankmRNA CR591143.1 - 1 262 Chr. 22 - 24573938 - 24574894 [+]
GenBankmRNA AV718073.1 - 1 282 Chr. 22 - 24573918 - 24574599 [+]
GenBankmRNA BX432338.2 - 1 262 Chr. 22 - 24573938 - 24574894 [+]
GenBankmRNA CN414448.1 - 1 17 Chr. 22 - 24574183 - 24574831 [+]
GenBankmRNA BX432339.2 - 1 262 Chr. 22 - 24573938 - 24574752 [+]

View Full Product Details