Gene Transcripts
- Gene Symbol
- NAG20
- Entrez Gene ID
- 100129703
- Gene Name
- NAG20
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.664650
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF210649.1 |
- |
1 |
240 |
|
View Full Product Details