Gene Transcripts
- Gene Symbol
- KCNK15
- Entrez Gene ID
- 60598
- Gene Name
- potassium two pore domain channel subfamily K member 15
- Gene Aliases
- K2p15.1, KCNK11, KCNK14, KT3.3, TASK-5, TASK5, dJ781B1.1
- Chromosome Location
- Chr. 20: 44745605 - 44752313 on Build GRCh38
- UniGene ID
- Hs.528664
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM049330.1 |
- |
1 |
572 |
|
Chr. 20 - 44750995 - 44751718 [+] |
View Full Product Details