Gene Transcripts
- Gene Symbol
- OSBPL1A
- Entrez Gene ID
- 114876
- Gene Name
- oxysterol binding protein like 1A
- Gene Aliases
- ORP-1, ORP1, OSBPL1B
- Chromosome Location
- Chr. 18: 24162045 - 24397882 on Build GRCh38
- UniGene ID
- Hs.370725
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021898.1 |
- |
1 |
716 |
|
View Full Product Details