Gene Transcripts
- Gene Symbol
- ICE1
- Entrez Gene ID
- 23379
- Gene Name
- interactor of little elongation complex ELL subunit 1
- Gene Aliases
- KIAA0947
- Chromosome Location
- Chr. 5: 5422673 - 5490234 on Build GRCh38
- UniGene ID
- Hs.449296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041002.2 |
- |
1 |
84 |
|
View Full Product Details