Gene Transcripts
- Gene Symbol
- HELLS
- Entrez Gene ID
- 3070
- Gene Name
- helicase, lymphoid-specific
- Gene Aliases
- ICF4, LSH, Nbla10143, PASG, SMARCA6
- Chromosome Location
- Chr. 10: 94545767 - 94613905 on Build GRCh38
- UniGene ID
- Hs.463677
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021443.1 |
- |
1 |
286 |
|
View Full Product Details