Gene Transcripts
- Gene Symbol
- SKCG-1
- Entrez Gene ID
- 100049615
- Gene Name
- sporadic kidney cancer gene 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.551193
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY662656.1 |
- |
1 |
491 |
|
View Full Product Details