Gene Transcripts
- Gene Symbol
- WRB
- Entrez Gene ID
- 7485
- Gene Name
- tryptophan rich basic protein
- Gene Aliases
- CHD5, GET1
- Chromosome Location
- Chr. 21: 39380287 - 39397889 on Build GRCh38
- UniGene ID
- Hs.198308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK293866.1 |
- |
5 |
636 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK125019.1 |
- |
1 |
359 |
|
Chr. 21 - 39406357 - 39409340 [+] |
View Full Product Details