Gene Transcripts
- Gene Symbol
- LINC01560
- Entrez Gene ID
- 203414
- Gene Name
- long intergenic non-protein coding RNA 1560
- Gene Aliases
- CXorf24
- Chromosome Location
- Chr. X: 47482716 - 47485227 on Build GRCh38
- UniGene ID
- Hs.597617
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126059.1 |
|
1 |
1092 |
|
GenBank mRNA |
BC025179.1 |
- |
1 |
237 |
|
HY147632.1 |
- |
1 |
296 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI260296.1 |
- |
1 |
237 |
|
Chr. X - 47483571 - 47484361 [+] |
View Full Product Details