Gene Transcripts
- Gene Symbol
- TIMM17B
- Entrez Gene ID
- 10245
- Gene Name
- translocase of inner mitochondrial membrane 17 homolog B (yeast)
- Gene Aliases
- DXS9822, JM3, TIM17B
- Chromosome Location
- Chr. X: 48893447 - 48898143 on Build GRCh38
- UniGene ID
- Hs.30570
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC029446.1 |
- |
1 |
302 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG542570.1 |
- |
1 |
303 |
|
Chr. X - 48897158 - 48897781 [-] |
View Full Product Details