Gene Transcripts
- Gene Symbol
- TNS3
- Entrez Gene ID
- 64759
- Gene Name
- tensin 3
- Gene Aliases
- TEM6, TENS1
- Chromosome Location
- Chr. 7: 47275154 - 47582553 on Build GRCh38
- UniGene ID
- Hs.520814
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL137468.1 |
- |
1 |
1207 |
|
View Full Product Details