Gene Transcripts
- Gene Symbol
- ZBTB46-AS1
- Entrez Gene ID
- 101928604
- Gene Name
- ZBTB46 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 20: 63808076 - 63823015 on Build GRCh38
- UniGene ID
- Hs.676054
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BI089231.1 |
- |
3 |
413 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK000832.1 |
- |
2 |
696 |
|
Chr. 20 - 63815596 - 63817423 [+] |
View Full Product Details