Gene Transcripts

Gene Symbol
ENSA
Entrez Gene ID
2029
Gene Name
endosulfine alpha
Gene Aliases
ARPP-19e
Chromosome Location
Chr. 1: 150622123 - 150629622 on Build GRCh38
UniGene ID
Hs.632456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK057673.1 - 1 695

View Full Product Details