Gene Transcripts
- Gene Symbol
- BICC1
- Entrez Gene ID
- 80114
- Gene Name
- BicC family RNA binding protein 1
- Gene Aliases
- BICC, CYSRD
- Chromosome Location
- Chr. 10: 58512970 - 58831437 on Build GRCh38
- UniGene ID
- Hs.100261
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA571439.1 |
- |
1 |
203 |
|
Chr. 10 - 58829878 - 58830710 [+] |
GenBankmRNA |
AA401855.1 |
- |
1 |
382 |
|
Chr. 10 - 58829699 - 58830341 [+] |
View Full Product Details