Gene Transcripts
- Gene Symbol
- LOC286191
- Entrez Gene ID
- 286191
- Gene Name
- uncharacterized LOC286191
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.720094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833212.1 |
- |
1 |
13 |
|
- Gene Symbol
- SBSPON
- Entrez Gene ID
- 157869
- Gene Name
- somatomedin B and thrombospondin type 1 domain containing
- Gene Aliases
- C8orf84, RPESP
- Chromosome Location
- Chr. 8: 73063077 - 73093272 on Build GRCh38
- UniGene ID
- Hs.439040
- Species
- Homo sapiens
View Full Product Details