Gene Transcripts
- Gene Symbol
- C16orf72
- Entrez Gene ID
- 29035
- Gene Name
- chromosome 16 open reading frame 72
- Gene Aliases
- PRO0149
- Chromosome Location
- Chr. 16: 9091329 - 9119698 on Build GRCh38
- UniGene ID
- Hs.221497
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094351.1 |
- |
1 |
990 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD523126.1 |
- |
1 |
388 |
|
Chr. 16 - 9105452 - 9106160 [+] |
View Full Product Details