Gene Transcripts

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
Chromosome Location
Chr. 12: 110614027 - 110649130 on Build GRCh38
UniGene ID
Hs.211511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC030993.1 - 1 428


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG399158.1 - 1 427 Chr. 12 - 110632450 - 110633076 [+]

View Full Product Details