Gene Transcripts
- Gene Symbol
- ATP6V1D
- Entrez Gene ID
- 51382
- Gene Name
- ATPase H+ transporting V1 subunit D
- Gene Aliases
- ATP6M, VATD, VMA8
- Chromosome Location
- Chr. 14: 67337864 - 67360003 on Build GRCh38
- UniGene ID
- Hs.272630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001155.1 |
- |
10 |
1221 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK125287.1 |
- |
1 |
982 |
|
Chr. 14 - 67334382 - 67336999 [-] |
View Full Product Details