Gene Transcripts
- Gene Symbol
- CDC42BPA
- Entrez Gene ID
- 8476
- Gene Name
- CDC42 binding protein kinase alpha
- Gene Aliases
- MRCK, MRCKA, PK428
- Chromosome Location
- Chr. 1: 226989865 - 227318492 on Build GRCh38
- UniGene ID
- Hs.35433
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130237.1 |
- |
1 |
646 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM699643.1 |
- |
1 |
78 |
|
Chr. 1 - 227060152 - 227060680 [-] |
View Full Product Details