Gene Transcripts
- Gene Symbol
- NOS1
- Entrez Gene ID
- 4842
- Gene Name
- nitric oxide synthase 1
- Gene Aliases
- IHPS1, N-NOS, NC-NOS, NOS, bNOS, nNOS
- Chromosome Location
- Chr. 12: 117208142 - 117441843 on Build GRCh38
- UniGene ID
- Hs.654410
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF446131.1 |
- |
1 |
412 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE297602.1 |
- |
1 |
359 |
|
Chr. 12 - 117331433 - 117452170 [-] |
View Full Product Details