Gene Transcripts
- Gene Symbol
- ARL13B
- Entrez Gene ID
- 200894
- Gene Name
- ADP ribosylation factor like GTPase 13B
- Gene Aliases
- ARL2L1, JBTS8
- Chromosome Location
- Chr. 3: 93980139 - 94055678 on Build GRCh38
- UniGene ID
- Hs.533086
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC073842.1 |
- |
1 |
567 |
|
BI821381.1 |
- |
1 |
566 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BP377404.1 |
- |
1 |
529 |
|
Chr. 3 - 93980161 - 93980732 [+] |
View Full Product Details