Gene Transcripts

Gene Symbol
SNRPD2P2
Entrez Gene ID
645339
Gene Name
small nuclear ribonucleoprotein D2 pseudogene 2
Gene Aliases
Chromosome Location
Chr. 1: 231475762 - 231476525 on Build GRCh38
UniGene ID
Hs.382049
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033826.1 1 245
GenBank mRNA BC014600.1 - 1 251


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG722592.1 - 1 251 Chr. 1 - 231475762 - 231476528 [-]

View Full Product Details