Gene Transcripts
- Gene Symbol
- HIVEP3
- Entrez Gene ID
- 59269
- Gene Name
- human immunodeficiency virus type I enhancer binding protein 3
- Gene Aliases
- KBP-1, KBP1, KRC, SHN3, Schnurri-3, ZAS3, ZNF40C
- Chromosome Location
- Chr. 1: 41506365 - 42035925 on Build GRCh38
- UniGene ID
- Hs.403972
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG199418.1 |
- |
3 |
594 |
|
Chr. 1 - 41847910 - 41918517 [-] |
GenBankmRNA |
BI823291.1 |
- |
3 |
790 |
|
Chr. 1 - 41848106 - 41918669 [-] |
View Full Product Details