Gene Transcripts
- Gene Symbol
- CDK6
- Entrez Gene ID
- 1021
- Gene Name
- cyclin dependent kinase 6
- Gene Aliases
- MCPH12, PLSTIRE
- Chromosome Location
- Chr. 7: 92604921 - 92836627 on Build GRCh38
- UniGene ID
- Hs.119882
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123134.1 |
- |
1 |
113 |
|
View Full Product Details