Gene Transcripts
- Gene Symbol
- RMST
- Entrez Gene ID
- 196475
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-protein coding)
- Gene Aliases
- LINC00054, NCRMS, NCRNA00054
- Chromosome Location
- Chr. 12: 97465021 - 97533766 on Build GRCh38
- UniGene ID
- Hs.652568
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF429306.1 |
- |
7 |
1308 |
|
View Full Product Details