Gene Transcripts
- Gene Symbol
- MORC3
- Entrez Gene ID
- 23515
- Gene Name
- MORC family CW-type zinc finger 3
- Gene Aliases
- NXP2, ZCW5, ZCWCC3
- Chromosome Location
- Chr. 21: 36320189 - 36376646 on Build GRCh38
- UniGene ID
- Hs.421150
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125011.1 |
- |
1 |
1556 |
|
View Full Product Details