Gene Transcripts
- Gene Symbol
- SFT2D1
- Entrez Gene ID
- 113402
- Gene Name
- SFT2 domain containing 1
- Gene Aliases
- C6orf83, pRGR1
- Chromosome Location
- Chr. 6: 166319728 - 166342606 on Build GRCh38
- UniGene ID
- Hs.487143
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090795.1 |
- |
1 |
119 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA145871.1 |
- |
1 |
119 |
|
Chr. 6 - 166321162 - 166321729 [-] |
View Full Product Details