Gene Transcripts
- Gene Symbol
- RUSC1-AS1
- Entrez Gene ID
- 284618
- Gene Name
- RUSC1 antisense RNA 1
- Gene Aliases
- C1orf104
- Chromosome Location
- Chr. 1: 155320460 - 155324147 on Build GRCh38
- UniGene ID
- Hs.650801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125510.1 |
- |
1 |
70 |
|
BC062571.1 |
- |
2 |
1533 |
5183308 |
View Full Product Details