Gene Transcripts
- Gene Symbol
- KRT7
- Entrez Gene ID
- 3855
- Gene Name
- keratin 7
- Gene Aliases
- CK7, K2C7, K7, SCL
- Chromosome Location
- Chr. 12: 52233170 - 52252667 on Build GRCh38
- UniGene ID
- Hs.411501
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042076.1 |
- |
1 |
71 |
|
View Full Product Details