Gene Transcripts
- Gene Symbol
- VN1R3
- Entrez Gene ID
- 317702
- Gene Name
- vomeronasal 1 receptor 3 (gene/pseudogene)
- Gene Aliases
- FKSG46, V1RL3, V1RL3p
- Chromosome Location
- Chr. 16: 31807590 - 31808846 on Build GRCh38
- UniGene ID
- Hs.257746
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF336873.1 |
- |
1 |
1110 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF333762.1 |
- |
|
584 |
|
Chr. - - [-] |
View Full Product Details