Gene Transcripts
- Gene Symbol
- XPNPEP3
- Entrez Gene ID
- 63929
- Gene Name
- X-prolyl aminopeptidase 3
- Gene Aliases
- APP3, ICP55, NPHPL1
- Chromosome Location
- Chr. 22: 40857081 - 40932819 on Build GRCh38
- UniGene ID
- Hs.529163
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC037424.2 |
- |
1 |
848 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB546196.1 |
- |
1 |
242 |
|
Chr. 22 - 40857697 - 40858025 [+] |
GenBankmRNA |
CB409178.1 |
- |
1 |
516 |
|
Chr. 22 - 40857423 - 40858013 [+] |
GenBankmRNA |
BM466568.1 |
- |
1 |
728 |
|
Chr. 22 - 40857233 - 40858036 [+] |
GenBankmRNA |
CB409437.1 |
- |
1 |
516 |
|
Chr. 22 - 40857423 - 40858004 [+] |
View Full Product Details