Gene Transcripts

Gene Symbol
IRAK1BP1
Entrez Gene ID
134728
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
AIP70, SIMPL
Chromosome Location
Chr. 6: 78867486 - 78979411 on Build GRCh38
UniGene ID
Hs.656212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010844.3 NP_001010844.1 4 1755
GenBank mRNA AK096861.1 - 1 427

View Full Product Details