Gene Transcripts
- Gene Symbol
- IRAK1BP1
- Entrez Gene ID
- 134728
- Gene Name
- interleukin 1 receptor associated kinase 1 binding protein 1
- Gene Aliases
- AIP70, SIMPL
- Chromosome Location
- Chr. 6: 78867486 - 78979411 on Build GRCh38
- UniGene ID
- Hs.656212
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM822771.1 |
- |
1 |
484 |
|
Chr. 6 - 78899889 - 78900508 [+] |
GenBankmRNA |
AA405600.1 |
- |
1 |
253 |
|
Chr. 6 - 78900121 - 78900548 [+] |
GenBankmRNA |
BX109533.1 |
- |
1 |
202 |
|
Chr. 6 - 78900172 - 78900966 [+] |
GenBankmRNA |
BX486475.1 |
- |
1 |
320 |
|
Chr. 6 - 78900056 - 78900649 [+] |
View Full Product Details