Gene Transcripts

Gene Symbol
DNAJB14
Entrez Gene ID
79982
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Gene Aliases
EGNR9427, PRO34683
Chromosome Location
Chr. 4: 99896248 - 99946726 on Build GRCh38
UniGene ID
Hs.577426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031723.3 NP_001026893.1 8 3942
NM_001278310.1 NP_001265239.1 7 3770
XM_011532262.2 XP_011530564.1 10 3826
XM_011532263.2 XP_011530565.1 8 4326
XM_011532265.2 XP_011530567.1 7 4270
XM_017008628.1 XP_016864117.1 10 4008
GenBank mRNA AK095074.1 - 1 26


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA307038.1 - 1 26 Chr. 4 - 99897828 - 99898406 [-]

View Full Product Details