Gene Transcripts

Gene Symbol
MYO9A
Entrez Gene ID
4649
Gene Name
myosin IXA
Gene Aliases
Chromosome Location
Chr. 15: 71822290 - 72118577 on Build GRCh38
UniGene ID
Hs.513000
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006720539.2 XP_006720602.1 43 12204
XM_011521613.2 XP_011519915.1 44 12261
XM_011521614.2 XP_011519916.1 45 11947
XM_011521615.2 XP_011519917.1 44 12114
XM_011521616.2 XP_011519918.1 44 11908
XM_011521617.2 XP_011519919.1 44 12258
XM_011521618.2 XP_011519920.1 43 12207
XM_011521619.2 XP_011519921.1 43 12204
XM_011521620.2 XP_011519922.1 43 12201
XM_011521621.2 XP_011519923.1 43 12141
XM_011521622.2 XP_011519924.1 43 12045
XM_011521623.2 XP_011519925.1 27 9364
XM_017022228.1 XP_016877717.1 44 11890
XM_017022229.1 XP_016877718.1 44 11880
XM_017022230.1 XP_016877719.1 42 11988
GenBank mRNA BC039397.1 - 1 896


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AW972853.1 - 134 Chr. - - [-]

View Full Product Details