Gene Transcripts
- Gene Symbol
- ABCC12
- Entrez Gene ID
- 94160
- Gene Name
- ATP binding cassette subfamily C member 12
- Gene Aliases
- MRP9
- Chromosome Location
- Chr. 16: 48082973 - 48146770 on Build GRCh38
- UniGene ID
- Hs.410111
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127951.1 |
- |
1 |
192 |
|
View Full Product Details