Gene Transcripts

Gene Symbol
TUSC3
Entrez Gene ID
7991
Gene Name
tumor suppressor candidate 3
Gene Aliases
D8S1992, M33, MRT22, MRT7, N33, OST3A
Chromosome Location
Chr. 8: 15540087 - 15766649 on Build GRCh38
UniGene ID
Hs.426324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017013861.1 XP_016869350.1 10 4861
GenBank mRNA BX641112.1 - 1 3064

View Full Product Details