Gene Transcripts
- Gene Symbol
- COX11
- Entrez Gene ID
- 1353
- Gene Name
- COX11, cytochrome c oxidase copper chaperone
- Gene Aliases
- COX11P
- Chromosome Location
- Chr. 17: 54951898 - 54971024 on Build GRCh38
- UniGene ID
- Hs.591171
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124809.1 |
- |
1 |
1385 |
|
View Full Product Details