Gene Transcripts

Gene Symbol
ZBTB7C
Entrez Gene ID
201501
Gene Name
zinc finger and BTB domain containing 7C
Gene Aliases
APM-1, APM1, ZBTB36, ZNF857C
Chromosome Location
Chr. 18: 48026886 - 48412639 on Build GRCh38
UniGene ID
Hs.515388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005258229.4 XP_005258286.1 4 4401
XM_011525861.2 XP_011524163.1 7 4881
XM_011525863.2 XP_011524165.1 6 4998
XM_011525864.2 XP_011524166.1 6 4753
XM_011525865.2 XP_011524167.1 6 4994
XM_011525866.2 XP_011524168.1 5 4739
XM_011525867.2 XP_011524169.1 5 4528
XM_011525869.2 XP_011524171.1 6 5003
XM_011525870.2 XP_011524172.1 5 4699
XM_011525871.2 XP_011524173.1 3 5214
XM_017025605.1 XP_016881094.1 7 6676
XM_017025606.1 XP_016881095.1 7 5130
XM_017025607.1 XP_016881096.1 7 4885
XM_017025608.1 XP_016881097.1 6 6553
XM_017025609.1 XP_016881098.1 5 4870
GenBank mRNA AK095226.1 - 2 2035

View Full Product Details