Gene Transcripts
- Gene Symbol
- ERO1A
- Entrez Gene ID
- 30001
- Gene Name
- endoplasmic reticulum oxidoreductase 1 alpha
- Gene Aliases
- ERO1-L, ERO1-L-alpha, ERO1-alpha, ERO1L, ERO1LA, Ero1alpha
- Chromosome Location
- Chr. 14: 52639915 - 52695931 on Build GRCh38
- UniGene ID
- Hs.525339
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM761643.1 |
- |
1 |
599 |
|
Chr. 14 - 52640527 - 52641194 [-] |
GenBankmRNA |
BM753238.1 |
- |
1 |
347 |
|
Chr. 14 - 52640381 - 52640942 [-] |
GenBankmRNA |
CR743961.1 |
- |
1 |
315 |
|
Chr. 14 - 52640408 - 52640910 [-] |
GenBankmRNA |
BM753824.1 |
- |
1 |
346 |
|
Chr. 14 - 52640410 - 52640942 [-] |
View Full Product Details